Learn More
Fluidigm Corp Maxpar PBS, 500 mL
SDP
Catalog No. NC1439049
Encompass
Formulated to provide the best possible signal to noise ratio for detection by mass cytometry.
Catalog No. NC1439049 Supplier Fluidigm Corp Supplier No. 201058
Formulated to provide the best possible signal to noise ratio for detection by mass cytometry.