Learn More
Fluidigm Corp MAXPAR X8 ANTIBODY LABEL KIT
SDP
Catalog No. NC0245957
Encompass
This Maxpar kit lets you conjugate IgG antibodies with the 170Er metal isotope, with each kit containing enough material to conjugate 440 antibodies in 100 g amounts.
Catalog No. NC0245957 Supplier Fluidigm Corp Supplier No. 201170A
This Maxpar kit lets you conjugate IgG antibodies with the 170Er metal isotope, with each kit containing enough material to conjugate 440 antibodies in 100 g amounts.