Learn More
Fluidigm Corp MAXPAR X8 ANTIBODY LABELING KI
SDP
Catalog No. NC0353812
Encompass
This Maxpar kit lets you conjugate IgG antibodies with the 156Gd metal isotope, with each kit containing enough material to conjugate 440 antibodies in 100 g amounts.
Catalog No. NC0353812 Supplier Fluidigm Corp Supplier No. 201156A
This Maxpar kit lets you conjugate IgG antibodies with the 156Gd metal isotope, with each kit containing enough material to conjugate 440 antibodies in 100 g amounts.