Learn More
Thermo Scientific™ Accessories for MaxQ™ 5000 Shakers
Catalog No. 11675461 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Gassing Manifold w/ 16 Outlets
Gassing Manifold w/ 8 Outlets
Shelf
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
11-675-461 Gassing Manifold w/ 8 Outlets
11-675-459 Gassing Manifold w/ 16 Outlets
11-675-460 Shelf
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-675-461 Supplier Thermo Scientific™ Supplier No. SHK50008GM
Add to Cart
Add to Cart
Increase the utility of the Thermo Scientific™ MaxQ™ 5000 Shaker with the MaxQ 5000 Gassing Manifold and Stainless-Steel Shelf.
Increase the utility of the Thermo Scientific™ MaxQ™ 5000 Shaker with the MaxQ 5000 Gassing Manifold and Stainless-Steel Shelf.
Gassing Manifold
  • Enables direct gassing into flasks for anaerobic cultures
  • Offered with 8 or 16 outlets
  • Easy-access drain system offers convenience in draining and disinfecting
Stainless-Steel Shelf
  • For back wall of chamber
  • L × W: 33.25 × 4.0″ (84.5 × 10.2cm)
  • Retractable foot casters allow easy manipulation in lab
TRUSTED_SUSTAINABILITY
Specifications
Product Type Gassing Manifold w/ 8 Outlets
For Use With (Equipment) MaxQ 5000