Learn More
Perkin Elmer US LLC Gentamicin ELISA Kit
Supplier:  Perkin Elmer US LLC FOOD102701A
Encompass_Preferred
The MaxSignal Gentamicin ELISA Kit is a competitive enzyme immunoassay for the quantitative analysis of gentamicin in milk, meat, liver, kidney, egg, embryonic egg, urine, serum, feed, honey, and soybean casein digest broth.
Catalog No. NC1837252