Learn More
Electron Microscopy Sciences Maxtaform Specialist Grid Style H9 Ni
SDP
Supplier:  Electron Microscopy Sciences H9SPECNI
Encompass_Preferred
High Grade Maxtaform grids with clean and smooth edges, firm support, and a large open area. These grids fill all of your special needs. 3 mm diameter, 0.75 mil thickness.
Catalog No. 50-287-83