Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jackson Safety Maxview™ Series Face Shield - Ratcheting - Chin and Side Guard

Catalog No. 19060842
Encompass
Encompass_Preferred
Change view
Click to view available options
Lens Coating:
Anti-fog
Uncoated
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating
19-060-842 Uncoated
19-060-843 Anti-fog
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-060-842 Supplier Jackson Safety Supplier No. 14200
Add to Cart
Add to Cart
This item is not returnable. View return policy

The MAXVIEW is a premium face shield that features an oversized window that delivers exceptional optical clarity and panoramic view. An integrated side and chin guard protects against splashes and flying debris while the extended crown protects against hazards from above.

  • Extra-large stylish crown and wraparound face shield frame
  • High-impact polycarbonate window (with or without anti-fog)

Specifications

Lens Coating Uncoated
Shape Contoured
Width (Metric) 24.45 cm
Height (English) 13 in.
Height (Metric) 33.02 cm
Width (English) 9.63 in.
Certifications/Compliance ANSI Z87.1 +high impact
Certifications/Compliance (Europe) CE Certified
Lens Tint Clear
Product Type Faceshield
Frame Color Black
Length (English) 9 in.
Length (Metric) 22.86 cm
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection None
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.