Learn More
Active Motif MAXwash™ Washing Medium
SDP
Catalog No. NC1841524
Encompass
MAXwash Washing Medium is a proprietary formulation designed and optimized to eliminate non-specific primary and secondary antibody binding. It has been tested and proven effective at eliminating off-target antibody binding in immunofluorescence assays.
Catalog No. NC1841524 Supplier Active Motif Supplier No. 15254
MAXwash Washing Medium is a proprietary formulation designed and optimized to eliminate non-specific primary and secondary antibody binding. It has been tested and proven effective at eliminating off-target antibody binding in immunofluorescence assays.