Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma-Aldrich May-Grunwald Stain

Catalog No. 01002215 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-215 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-215 Supplier Sigma-Aldrich Supplier No. MG1284L
Add to Cart
Add to Cart

Stain for use in the differential staining of cellular elements of blood.

  • For hematology and histology applications
TRUSTED_SUSTAINABILITY

Specifications

Quantity 4 L
Storage Requirements Room Temperature
Format Liquid
Concentration 0.25% (w/v) in Methanol
For Use With (Application) in vitro Diagnostic Use
Product Type May-Grunwald Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.