Learn More
May-Grunwald Stain, Sigma-Aldrich
Catalog No. 01259007 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
Catalog No. Quantity
01-259-007 25 g
1 options
Catalog No. 01-259-007 Supplier MilliporeSigma Supelco Supplier No. 20543525G
Add to Cart
Add to Cart
Suitable for the differential staining of cellular elements of blood.
May-Grunwald stain is a Romanowsky neutraldye stain based on the combination of methylene blue azures as the basic dyecomponent and eosin as the acid dyecomponent. It is intended for use in staining blood films orbone marrow films. It is used routinely for diagnostic purposes in hematology.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 g
Product Type May-Grunwald Solution