Learn More
Electron Microscopy Sciences Mayer's Hematoxylin, 500L
SDP
Catalog No. 5027048
Encompass_Preferred
Mayer's Hematoxylin is used as a counterstain for procedures such as immunohistochemistry or laser microdissection.
Catalog No. 50-270-48 Supplier Electron Microscopy Sciences Supplier No. 2604305
Mayer's Hematoxylin is used as a counterstain for procedures such as immunohistochemistry or laser microdissection.