Learn More
Mayer's Hematoxylin Solution, Sigma-Aldrich
Catalog No. 01259005 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-005 500 mL
01-258-987 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-259-005 Supplier MilliporeSigma Supelco Supplier No. 51275500ML
Add to Cart
Add to Cart
An alcohol-free nuclear counterstain used for histological techniques that preserves stained cytoplasmic components.
Selective staining with hematoxylin, reviewcitation. For staining nuclear materialcitation
Mayer's Hematoxylin Solution, Sigma-Aldrich, chemical
TRUSTED_SUSTAINABILITY
Specifications
Quantity 500 mL
Product Type Mayers Hematoxylin Stain