Learn More
Electron Microscopy Sciences Mayers Hematoxylin 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2625201
Encompass_Preferred
Used in Maximow's Method for Bone Marrow, Morphology details of marrow cells. Mallory (1961). Morphology details of marrow cells. See AFIP Manual of Histologic Staining Methods, 3rd Ed, Ed. L. Luna: New York: McGraw Hill Publications, c. 1968, p. 120 or Mallory, F.B., Pathological technique, New York, Hafner Publishing Co., 1961, p.196
Catalog No. 50-321-35