Learn More
Electron Microscopy Sciences Mayers Hematoxylin 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2660202
Encompass_Preferred
Used in Rhodanine Method for Copper, Mineral and Pigment stain. See Sheehan and Hrapchack, Theory and Practice of Histotechnology. St. Louis, The Mosby Company 1980 p. 230.
Catalog No. 50-318-51