Learn More
Fine Science Tools Mayo Scissors, Tungsten Carbide, 15 cm
SDP
Catalog No. NC9851195
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 39mm/ Alloy / Material: Stainless Steel/ Length: 15cm/ Serrated: No/ Feature: Tungsten Carbide
Catalog No. NC9851195 Supplier Fine Science Tools Supplier No. 1451015
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 39mm/ Alloy / Material: Stainless Steel/ Length: 15cm/ Serrated: No/ Feature: Tungsten Carbide