Learn More
Roboz Surgical Roboz Surgical Mayo Scissors, 5.5 Inch, Straight, 1 ea
SDP
Catalog No. NC9775231
Encompass
These 5.5 in. straight Mayo scissors are precision-engineered surgical instruments designed for cutting dense tissues such as fascia or thick muscles. Featuring a robust, durable construction, they are suitable for a variety of general surgical procedures requiring reliable, clean cutting performance.
  • Straight design for versatile surgical access
  • Total instrument length of 5.5 in.
  • Blade length of 43 mm
  • Constructed for repeated use in demanding surgical environments
Catalog No. NC9775231 Supplier Roboz Surgical Supplier No. RS6870
These 5.5 in. straight Mayo scissors are precision-engineered surgical instruments designed for cutting dense tissues such as fascia or thick muscles. Featuring a robust, durable construction, they are suitable for a variety of general surgical procedures requiring reliable, clean cutting performance.
  • Straight design for versatile surgical access
  • Total instrument length of 5.5 in.
  • Blade length of 43 mm
  • Constructed for repeated use in demanding surgical environments