Learn More
Sklar Instruments Surgi-OR Mayo Dissecting Scissors, Sklar 5-1/2 in., Blunt/Blunt, Straight, Smooth, Finger Rings, Stainless Steel. EA.
SDP
Catalog No. 501184194 Shop All Sklar Instruments Products
Encompass_Preferred
Surgi-OR Mayo Dissecting Scissors, Sklar. 5-1/2 in., Blunt/Blunt, Straight, Smooth, Finger Rings, Stainless Steel. Surgi-OR Mayo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and multiple settings. They are available straight or curved in a variety of lengths with a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. This product is straight with smooth, blunt/blunt tips and a length of 5-1/2 inches.
Catalog No. 50-118-4194 Supplier Sklar Instruments Supplier No. 95308
Surgi-OR Mayo Dissecting Scissors, Sklar. 5-1/2 in., Blunt/Blunt, Straight, Smooth, Finger Rings, Stainless Steel. Surgi-OR Mayo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and multiple settings. They are available straight or curved in a variety of lengths with a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. This product is straight with smooth, blunt/blunt tips and a length of 5-1/2 inches.