Learn More
Roboz Surgical MAYO DISSECTNG SCISSOR 5-1/2IN
SDP
Catalog No. NC0092069
Encompass
These straight Mayo scissors are precision-engineered surgical instruments designed for reliable tissue dissection. Featuring durable tungsten carbide inserts, these scissors maintain a sharp, long-lasting edge for consistent performance in demanding surgical environments.
  • Tungsten carbide blades for enhanced durability and edge retention
  • Straight configuration suitable for a variety of general dissection tasks
  • Overall length of 5.5 in. provides balanced control
  • Blade length of 43 mm optimized for precise cutting
Catalog No. NC0092069 Supplier Roboz Surgical Supplier No. RS6880
These straight Mayo scissors are precision-engineered surgical instruments designed for reliable tissue dissection. Featuring durable tungsten carbide inserts, these scissors maintain a sharp, long-lasting edge for consistent performance in demanding surgical environments.
  • Tungsten carbide blades for enhanced durability and edge retention
  • Straight configuration suitable for a variety of general dissection tasks
  • Overall length of 5.5 in. provides balanced control
  • Blade length of 43 mm optimized for precise cutting