Learn More
Med Vet International Miltex Vantage Mayo-Hegar Needle Holder, 8in.
Catalog No. 502831215
Encompass_Preferred
The Miltex Vantage V98-48 Mayo-Hegar needle holder is made from high quality Japanese stainless steel and measures 8" in length. It has a finger ring handle with a ratchet lock and straight, serrated jaws.
Catalog No. 50-283-1215 Supplier Med Vet International Supplier No. MILV9848
The Miltex Vantage V98-48 Mayo-Hegar needle holder is made from high quality Japanese stainless steel and measures 8" in length. It has a finger ring handle with a ratchet lock and straight, serrated jaws.