Learn More
Sklar Instruments Mayo-Hegar Needle Holder, 6 in., Straight, Cross-Serrated with Groove, Stainless Steel, EA.
SDP
Catalog No. 501109760 Shop All Sklar Instruments Products
Encompass_Preferred
Mayo-Hegar Needle Holder, 6 in., Straight, Cross-Serrated with Groove, Stainless Steel, EA. The Surgi-OR Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures. Due to it's versatility, it is one of the most commonly used surgical tools for both human and animal procdures. Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths of this product may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is commonly used with larger, heavier suture needles because of it's wide jaws. Surgi-OR instruments are mid-grade instruments, suitable for use in the operating room or surgery center. This product is straight, has cross-serrated jaws featuring a central groove, and measures 6 inches overall.
Catalog No. 50-110-9760 Supplier Sklar Instruments Supplier No. 95847
Mayo-Hegar Needle Holder, 6 in., Straight, Cross-Serrated with Groove, Stainless Steel, EA. The Surgi-OR Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures. Due to it's versatility, it is one of the most commonly used surgical tools for both human and animal procdures. Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths of this product may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is commonly used with larger, heavier suture needles because of it's wide jaws. Surgi-OR instruments are mid-grade instruments, suitable for use in the operating room or surgery center. This product is straight, has cross-serrated jaws featuring a central groove, and measures 6 inches overall.