Learn More
Sklar Instruments Mayo-Hegar Needle Holder, Sklar, 10-1/2 in., Blunt, Straight, Cross-Serrated, Finger Rings, Stainless Steel. EA/1.
SDP
Catalog No. 501437492 Shop All Sklar Instruments Products
Encompass_Preferred
Mayo-Hegar Needle Holder, Sklar, 10-1/2 in., Blunt, Straight, Cross-Serrated, Finger Rings, Stainless Steel. The Sklar Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures. Due to it's versatility, it is one of the most commonly used surgical tools for both human and animal procdures, typically with larger, heavier suture needles because of it's wide jaws. Shorter lengths of the instrument may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. This product is straight with cross-serrated jaws featuring a central groove where the needle can be held, and a length of 10-1/2 inches. This product may also be known as a Masson needleholder.
This item is not returnable. View return policy
Catalog No. 50-143-7492 Supplier Sklar Instruments Supplier No. 202910
Mayo-Hegar Needle Holder, Sklar, 10-1/2 in., Blunt, Straight, Cross-Serrated, Finger Rings, Stainless Steel. The Sklar Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures. Due to it's versatility, it is one of the most commonly used surgical tools for both human and animal procdures, typically with larger, heavier suture needles because of it's wide jaws. Shorter lengths of the instrument may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. This product is straight with cross-serrated jaws featuring a central groove where the needle can be held, and a length of 10-1/2 inches. This product may also be known as a Masson needleholder.