Learn More
Sklar Instruments Mayo-Hegar Needle Holder, Sklar, 5-1/4 in., Blunt, Straight, Cross-Serrated with Groove, Stainless Steel. EA/1.
SDP
Catalog No. 501437468 Shop All Sklar Instruments Products
Encompass_Preferred
Mayo-Hegar Needle Holder, Sklar, 5-1/4 in., Blunt, Straight, Cross-Serrated with Groove, Stainless Steel. The Sklar Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures. Due to it's versatility, it is one of the most commonly used surgical tools for both human and animal procdures. Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths of this product may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is commonly used with larger, heavier suture needles because of it's wide jaws. This product is straight with cross-serrated jaws featuring a central groove with which the needle can be held, and a length of 5-1/4 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7468 Supplier Sklar Instruments Supplier No. 202052
Mayo-Hegar Needle Holder, Sklar, 5-1/4 in., Blunt, Straight, Cross-Serrated with Groove, Stainless Steel. The Sklar Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures. Due to it's versatility, it is one of the most commonly used surgical tools for both human and animal procdures. Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths of this product may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is commonly used with larger, heavier suture needles because of it's wide jaws. This product is straight with cross-serrated jaws featuring a central groove with which the needle can be held, and a length of 5-1/4 inches.