Learn More
Med Vet International Mayo-Hegar Needle Holder, 6-in. (15.24cm), German-Made Tungsten Carbide, J0078WTA
Catalog No. 5000584827
Encompass_Preferred
Needle Holders featured on our site are made with special tungsten carbide jaws. Tung- sten carbide will hold tight enough that even round-bodied needles don’t twist or turn. • Jaw edges are chamfered to prevent accidentally cutting suture• Finger rings are well rounded and polished for better feel and prevention of finger fatigue• Satin finish to reduce glare• Gold plated finger rings for easy identificationGerman-Made. Tungsten carbide jaws. 6"
Catalog No. 50-005-84827 Supplier Med Vet International Supplier No. JOR78WTA
Needle Holders featured on our site are made with special tungsten carbide jaws. Tung- sten carbide will hold tight enough that even round-bodied needles don’t twist or turn. • Jaw edges are chamfered to prevent accidentally cutting suture• Finger rings are well rounded and polished for better feel and prevention of finger fatigue• Satin finish to reduce glare• Gold plated finger rings for easy identificationGerman-Made. Tungsten carbide jaws. 6"