Learn More
Med Vet International Mayo-Hegar Needle Holder, 7-in. (17.78cm) Tungsten Carbide Jaws, German-Made, J0078WTB
Catalog No. 5000584852
Encompass_Preferred
Needle Holders featured on our site are made with special tungsten carbide jaws. Tung- sten carbide will hold tight enough that even round-bodied needles don’t twist or turn. • Jaw edges are chamfered to prevent accidentally cut- ting suture• Finger rings are well rounded and polished for better feel and prevention of finger fatigue• Satin finish to reduce glare• Gold plated finger rings for easy identificationGerman-Made. Tungsten carbide jaws. 7"
Catalog No. 50-005-84852 Supplier Med Vet International Supplier No. JOR78WTB
Needle Holders featured on our site are made with special tungsten carbide jaws. Tung- sten carbide will hold tight enough that even round-bodied needles don’t twist or turn. • Jaw edges are chamfered to prevent accidentally cut- ting suture• Finger rings are well rounded and polished for better feel and prevention of finger fatigue• Satin finish to reduce glare• Gold plated finger rings for easy identificationGerman-Made. Tungsten carbide jaws. 7"