Learn More
Med Vet International Miltex Vantage Mayo Dissecting Scissors, 5.5in., Straight
Catalog No. 502832037
Encompass_Preferred
The Miltex Vantage V95-120 Mayo dissecting scissors are made from high quality Japanese stainless steel and measure 5.5" in length. They have a finger ring handle and straight blades with blunt tips.
Catalog No. 50-283-2037 Supplier Med Vet International Supplier No. MILV95120
The Miltex Vantage V95-120 Mayo dissecting scissors are made from high quality Japanese stainless steel and measure 5.5" in length. They have a finger ring handle and straight blades with blunt tips.