Learn More
Med Vet International Miltex Vantage Mayo Dissecting Scissors, 6.75in., Straight
Supplier:  Med Vet International MILV95124
Encompass_Preferred
The Miltex Vantage V95-124 Mayo dissecting scissors are made from high quality Japanese stainless steel and measure 6.75" in length. They have a finger ring handle and straight blades with blunt tips.
Catalog No. 50-283-2039