Learn More
Fine Science Tools MAYO SCISSORS
SDP
Catalog No. NC1163844
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 45mm/ Alloy / Material: Stainless Steel/ Length: 17cm/ Serrated: No/ Feature: Tungsten Carbide
Catalog No. NC1163844 Supplier Fine Science Tools Supplier No. 1451017
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 45mm/ Alloy / Material: Stainless Steel/ Length: 17cm/ Serrated: No/ Feature: Tungsten Carbide