Learn More
World Precision Instrument Mayo Scissors, 16cm, SuperCut, Curved
SDP
Catalog No. 502539513
Encompass
SuperCut Mayo Scissors, also called Mayo Dissecting Scissors, are surgical scissors made popular by the Mayo Clinic. They have ring handles and long, straight, thick blades with blunt tips that are straight or curved. The surgical scissors vary in length from 14 cm (5.5") to 25 cm (9.8"). Longer blades give you greater reach for deeper dissection. The blades are sturdy for heavy duty work like cutting connective tissues, fascia, and sutures. Mayo Scissors are often used for dissection, where the blunt tips prevent accidental damage to surrounding tissue and may be used to delicately probe. They may also be used for dressing and bandage removal.
Catalog No. 50-253-9513 Supplier World Precision Instrument Supplier No. 503278
SuperCut Mayo Scissors, also called Mayo Dissecting Scissors, are surgical scissors made popular by the Mayo Clinic. They have ring handles and long, straight, thick blades with blunt tips that are straight or curved. The surgical scissors vary in length from 14 cm (5.5") to 25 cm (9.8"). Longer blades give you greater reach for deeper dissection. The blades are sturdy for heavy duty work like cutting connective tissues, fascia, and sutures. Mayo Scissors are often used for dissection, where the blunt tips prevent accidental damage to surrounding tissue and may be used to delicately probe. They may also be used for dressing and bandage removal.