Learn More
Roboz Surgical Roboz Mayo Scissors, Curved, Sure Cut, 6.75 in, 1 ea
SDP
Catalog No. 5000574781
Encompass
These curved Mayo scissors are precision-engineered for clean, atraumatic tissue dissection. Featuring a 6 3/4-inch overall length and 53mm blade length, they incorporate specialized cutting surfaces designed to minimize tissue trauma and improve cutting efficiency.
  • One blade features perpendicular micro-serrations to grip soft or tough tissue and prevent slippage.
  • Opposing blade is honed to razor sharpness for a smooth, reliable cut with less required force.
  • Curved geometry allows for improved visibility and access during surgical procedures.
  • Black ring handles enable easy visual identification.
  • Designed specifically for tissue cutting applications.
Catalog No. 50-005-74781 Supplier Roboz Surgical Supplier No. RS6873SC
These curved Mayo scissors are precision-engineered for clean, atraumatic tissue dissection. Featuring a 6 3/4-inch overall length and 53mm blade length, they incorporate specialized cutting surfaces designed to minimize tissue trauma and improve cutting efficiency.
  • One blade features perpendicular micro-serrations to grip soft or tough tissue and prevent slippage.
  • Opposing blade is honed to razor sharpness for a smooth, reliable cut with less required force.
  • Curved geometry allows for improved visibility and access during surgical procedures.
  • Black ring handles enable easy visual identification.
  • Designed specifically for tissue cutting applications.