Learn More
Electron Microscopy Sciences EMS SuperCut Mayo Scissors with Straight Blades, 9"
SDP
Catalog No. 501931422
Encompass_Preferred
SuperCut scissors are made in Germany using the highest grade of surgical steel and are tempered and hardened for an extended and sharper life than ordinary steel. Rounded and polished finger rings Straight version cuts into surface area tissue.
Blade: Straight
OAL: 9"
This item is not returnable. View return policy
Catalog No. 50-193-1422 Supplier Electron Microscopy Sciences Supplier No. 7299615
SuperCut scissors are made in Germany using the highest grade of surgical steel and are tempered and hardened for an extended and sharper life than ordinary steel. Rounded and polished finger rings Straight version cuts into surface area tissue.
Blade: Straight
OAL: 9"