Learn More
Fine Science Tools MAYO STILLE SCISSORS
SDP
Catalog No. NC9557760
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 39mm/ Alloy / Material: Stainless Steel/ Length: 15cm/ Serrated: No
Catalog No. NC9557760 Supplier Fine Science Tools Supplier No. 1401215
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 39mm/ Alloy / Material: Stainless Steel/ Length: 15cm/ Serrated: No