Learn More
Stoelting™ Mayo-Stille Scissors
SDP
Catalog No. 10001714 Shop All Stoelting Co Products
Change view
Click to view available options
Length (English):
5.5 in.
6.75 in.
Model:
Curved
Straight
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Model
10-001-714 5.5 in. Straight
10-001-715 5.5 in. Curved
10-001-716 6.75 in. Straight
10-001-717 6.75 in. Curved
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10-001-714 Supplier Stoelting™ Supplier No. 5213664
Add to Cart
Add to Cart
Straight and curved scissors
  • Straight-blade style is designed for cutting body tissues near the surface of a wound.
  • Curved-blade style allows deeper penetration into the wound than the type with straight blades and is used to cut thick tissues
Specifications
For Use With (Application) Neuroscience
Length (English) 5.5 in.
Model Straight