Learn More
Sklar Instruments Mayo-Stille Dissecting Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Straight, Smooth. Stainless Steel. Sklar Mayo-Stille Dissecting Scissors have rounded edges which permits deep dissection of tough tissue. EA/1.
SDP
Catalog No. 501437367 Shop All Sklar Instruments Products
Encompass_Preferred
Mayo-Stille Dissecting Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Straight, Smooth. Stainless Steel. Sklar Mayo-Stille Dissecting Scissors have rounded edges which permits deep dissection of tough tissue. This product is straight with smooth, blunt/blunt tips, and an overall length of 5-1/2 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7367 Supplier Sklar Instruments Supplier No. 151655
Mayo-Stille Dissecting Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Straight, Smooth. Stainless Steel. Sklar Mayo-Stille Dissecting Scissors have rounded edges which permits deep dissection of tough tissue. This product is straight with smooth, blunt/blunt tips, and an overall length of 5-1/2 inches.