Learn More
Medchemexpress LLC Maytansine | 35846-53-8 | MFCD28127038 | 99.8% | C34H46ClN3O10 | 10MG
SDP
Catalog No. 5000261508
Encompass_Preferred
Maytansine is a highly potent microtubule-targeted natural product that induces mitotic arrest and kills tumor cells at subnanomolar concentrations. It is used as a research reagent to study microtubule dynamics and as a cytotoxic payload in antibody-drug conjugate (ADC) development.
  • Potent microtubule inhibitor active at subnanomolar concentrations.
  • Useful for cell biology and biochemical assays of microtubule dynamics.
  • Commonly used as a cytotoxic payload in ADC development and conjugation research.
  • High purity suitable for research use (99.8%).
  • Available in small milligram pack sizes for laboratory experiments.
Catalog No. 50-002-61508 Supplier Medchemexpress LLC Supplier No. HY1367410MG
Maytansine is a highly potent microtubule-targeted natural product that induces mitotic arrest and kills tumor cells at subnanomolar concentrations. It is used as a research reagent to study microtubule dynamics and as a cytotoxic payload in antibody-drug conjugate (ADC) development.
  • Potent microtubule inhibitor active at subnanomolar concentrations.
  • Useful for cell biology and biochemical assays of microtubule dynamics.
  • Commonly used as a cytotoxic payload in ADC development and conjugation research.
  • High purity suitable for research use (99.8%).
  • Available in small milligram pack sizes for laboratory experiments.