Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Maytansine, N2'-deacetyl-N2'-[27-(11,12-didehydrodibenz[b,f]azocin-5(6H)-yl)-2,3-dimethyl | 2479378-44-2 | MFCD02066234 | 94.4% | 1297.92 | C68H89ClN6O17 | 1 MG
SDP

Catalog No. 5000203439
Encompass_Preferred

DBCO-PEG4-Ahx-DM1 is a drug-linker conjugate that couples the cytotoxic maytansinoid DM1 to a DBCO-PEG4-Ahx clickable linker for use in antibody-drug conjugate development and conjugation chemistry.

  • Intended for antibody-drug conjugate (ADC) construction.
  • DBCO functional group enables copper-free click chemistry conjugation.
  • PEG4-Ahx spacer provides flexibility and improved solubility.
  • Contains DM1 maytansinoid as the cytotoxic payload.
  • High purity (≈94.4%).
  • Available in small research pack sizes and supplied as a solid.
  • Storage: powder -20 °C (stable for 3 years); in solvent -80 °C for 6 months or -20 °C for 1 month.

Catalog No. 50-002-03439 Supplier Medchemexpress LLC Supplier No. HY1362601MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-PEG4-Ahx-DM1 is a drug-linker conjugate that couples the cytotoxic maytansinoid DM1 to a DBCO-PEG4-Ahx clickable linker for use in antibody-drug conjugate development and conjugation chemistry.

  • Intended for antibody-drug conjugate (ADC) construction.
  • DBCO functional group enables copper-free click chemistry conjugation.
  • PEG4-Ahx spacer provides flexibility and improved solubility.
  • Contains DM1 maytansinoid as the cytotoxic payload.
  • High purity (≈94.4%).
  • Available in small research pack sizes and supplied as a solid.
  • Storage: powder -20 °C (stable for 3 years); in solvent -80 °C for 6 months or -20 °C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.