Learn More
Cayman Chemical Maytansinol, Purity: ≥95%, CAS: 57103-68-1, 100 mg
SDP
Catalog No. 502937258
Encompass_Preferred
An ansa macrolide; inhibits polymerization and induces depolymerization of bovine brain tubulin (EC50s = 12 and 43 µM, respectively); inhibits sea urchin egg mitosis at 10 µM and decreases proliferation of KB nasopharyngeal cancer cells (EC50 = 0.19 µg/ml)
Catalog No. 50-293-7258 Supplier Cayman Chemical Supplier No. 29228100 MG
An ansa macrolide; inhibits polymerization and induces depolymerization of bovine brain tubulin (EC50s = 12 and 43 µM, respectively); inhibits sea urchin egg mitosis at 10 µM and decreases proliferation of KB nasopharyngeal cancer cells (EC50 = 0.19 µg/ml)