Learn More
Cayman Chemical Maytnsnol 10mg
SDP
Catalog No. 502938114
Encompass_Preferred
An ansa macrolide; inhibits polymerization and induces depolymerization of bovine brain tubulin (EC50s = 12 and 43 µM, respectively); inhibits sea urchin egg mitosis at 10 µM and decreases proliferation of KB nasopharyngeal cancer cells (EC50 = 0.19 UG/ml)
Catalog No. 50-293-8114 Supplier Cayman Chemical Supplier No. 2922810 MG
An ansa macrolide; inhibits polymerization and induces depolymerization of bovine brain tubulin (EC50s = 12 and 43 µM, respectively); inhibits sea urchin egg mitosis at 10 µM and decreases proliferation of KB nasopharyngeal cancer cells (EC50 = 0.19 UG/ml)