Learn More
Aviva Systems Biology Corp MAZ Antibody - N-terminal region (ARP38146_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38146_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAZ. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MAZ. Alternate names: PUR1, Pur-1, SAF-1, SAF-2, ZF87, ZNF801, Zif87, SAF-3ApplicationIHC, WB. Species reactivity: Human, Mouse, Pig, Rat
Catalog No. 50-014-56536