Learn More
Medisca™ MAZ™ Counter Balance Weight Set
SDP
Catalog No. 14186477 Shop All Medisca Products
Change view
Click to view available options
For Use With (Equipment):
KK-400W
Catalog No. For Use With (Equipment)
14-186-477 KK-400W
1 options
Catalog No. 14-186-477 Supplier Medisca™ Supplier No. 611101
Add to Cart
Add to Cart
Counter balance weight set for use in compatible holders to facilitate balancing before cycles, with engraved weights for identification including 200 g, 100 g, 50 g, 40 g, 25 g, and 15 g.
  • Designed for use with the MAZ™ KK-400W holder to perform the required counterbalance step before operation
  • Can be placed directly into the holder for quick setup
  • Each weight has its mass engraved for easy identification
  • Includes: 1 x 200 g, 1 x 100 g, 1 x 50 g, 1 x 40 g, 1 x 25 g, 1 x 15 g
Specifications
Product Type Counter Balance
For Use With (Equipment) KK-400W
Includes 1 x 200 g, 1 x 100 g, 1 x 50 g, 1 x 40 g, 1 x 25 g, 1 x 15 g