Learn More
Medisca™ MAZ™ KK-250S and KK-300SS Standard Container Adapter Set
SDP
Catalog No. 14186476 Shop All Medisca Products
Change view
Click to view available options
Quantity:
2 Pc.
Catalog No. Quantity
14-186-476 2 Pc.
1 options
Catalog No. 14-186-476 Supplier Medisca™ Supplier No. 636901
Add to Cart
Add to Cart
Adapter set allows standard containers from KK-250S and KK-300SS to be used in KK-1000W mixers for flexible operation.
  • Compatible with KK-250 and KK-300SS standard containers
  • Designed for use with Mazerustar KK-1000W
  • Enables standard containers from KK-250 and KK-300SS to be used with the KK-1000W
  • Provides flexibility and compatibility across different models
Specifications
Product Type Contianer Adpater
Model KK-1000W
Quantity 2 Pc.