Learn More
Medisca™ MAZ™ Mixing Liner
SDP
Catalog No. 14186475 Shop All Medisca Products
Change view
Click to view available options
For Use With (Equipment):
KK-1000W
KK-300SS
KK-400W
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-186-475 KK-400W
14-186-474 KK-1000W
14-186-473 KK-300SS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-186-475 Supplier Medisca™ Supplier No. 621301
Add to Cart
Add to Cart
Stainless steel mixing liner for use with standard mixing container for micronizing purposes.
  • Made from 316L stainless steel for enhanced corrosion resistance
  • Designed to be used with the standard mixing container
  • Suitable for micronizing applications
  • Supports efficient processing of fine materials
Specifications
Product Type Stainless Steel Jar
Material Stainless Steel
Model KK-400W
For Use With (Equipment) KK-400W