Learn More
Medisca™ MAZ™ Zirconia Milling Beads
SDP
Catalog No. 14186472 Shop All Medisca Products
Change view
Click to view available options
For Use With (Equipment):
KK-300SS, KK-400W, KK-1000W, KK-2000W
Catalog No. For Use With (Equipment)
14-186-472 KK-300SS, KK-400W, KK-1000W, KK-2000W
1 options
Catalog No. 14-186-472 Supplier Medisca™ Supplier No. 641501
Add to Cart
Add to Cart
Durable zirconia milling beads for efficient ball milling and attrition, providing contamination free processing for pharmaceutical, food, chemical, and textile applications.
  • Highly durable milling beads designed for ball milling and attrition processes
  • Provides a contamination-free grinding solution
  • Suitable for pharmaceutical, food, chemical, and textile applications
Features:
  • Offers higher grinding efficiency with reduced processing time
  • Made from durable material, lowering long-term operational costs
  • Suitable for high-velocity and wet grinding operations
Specifications
Product Type Milling Beads
Material Zirconia
For Use With (Equipment) KK-300SS, KK-400W, KK-1000W, KK-2000W