Learn More
Medchemexpress LLC Mazdutide TFA | 2259884-03-0 | 99.7% | 4563.06 | 1 MG
SDP
Catalog No. 5000398693
Encompass_Preferred
Mazdutide TFA is a long-acting synthetic oxyntomodulin analog. It acts as a co-agonist of glucagon-like peptide (GLP-1R) and glucagon receptor (GCGR). It stimulates insulin secretion from mouse islets and is utilized in studies for obesity and type 2 diabetes (T2D).
  • Acts as a co-agonist of GLP-1R and GCGR
  • Stimulates insulin secretion from mouse islets
  • Used in studies of obesity and type 2 diabetes
  • Reduces body weight and physical intake in diabetic mice with diet-induced obesity
Catalog No. 50-003-98693 Supplier Medchemexpress LLC Supplier No. HYP3375A1MG
Mazdutide TFA is a long-acting synthetic oxyntomodulin analog. It acts as a co-agonist of glucagon-like peptide (GLP-1R) and glucagon receptor (GCGR). It stimulates insulin secretion from mouse islets and is utilized in studies for obesity and type 2 diabetes (T2D).
  • Acts as a co-agonist of GLP-1R and GCGR
  • Stimulates insulin secretion from mouse islets
  • Used in studies of obesity and type 2 diabetes
  • Reduces body weight and physical intake in diabetic mice with diet-induced obesity