Learn More
TARGETMOL CHEMICALS INC MB-07729 5MG
SDP
Catalog No. 502581499
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. MB-07729 is a potent non-competitive inhibitor of fructose-16-bisphosphate with IC50 values of 189 121 and 31 nM in rat monkey and human respectively for the study of diabetes. purity: 99%
Catalog No. 50-258-1499 Supplier TARGETMOL CHEMICALS INC Supplier No. T119555MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. MB-07729 is a potent non-competitive inhibitor of fructose-16-bisphosphate with IC50 values of 189 121 and 31 nM in rat monkey and human respectively for the study of diabetes. purity: 99%