Learn More
Quip Laboratories Inc MB-10 6G TABLETS 80/CS
SDP
Catalog No. NC0851290
Encompass
MB-10 Tablets are the most convenient way yet to deliver the benefits of chlorine dioxide to your facility hygiene and biosafety program. Simply drop this effervescent tablet into a pint, quart or gallon of water and, within minutes, the tablet dissolves into a safe, stable, multi-functional surface disinfectant and sterilant solution.
This item is not returnable. View return policy
Catalog No. NC0851290 Supplier Quip Laboratories Inc Supplier No. MBTAB80BX
MB-10 Tablets are the most convenient way yet to deliver the benefits of chlorine dioxide to your facility hygiene and biosafety program. Simply drop this effervescent tablet into a pint, quart or gallon of water and, within minutes, the tablet dissolves into a safe, stable, multi-functional surface disinfectant and sterilant solution.