Learn More
Selleck Chemical LLC MB 311-A2671-1MG
SDP
Catalog No. 503136684
Encompass_Preferred
MB 311 is a fully humanized monoclonal antibody recognizing the tumor-associated antigen Lewis Y It represents an attractive target for cancer immunotherapy and can be used in treatment of breast cancer MW 146 38 KD
Catalog No. 50-313-6684 Supplier Selleck Chemical LLC Supplier No. A26711MG
MB 311 is a fully humanized monoclonal antibody recognizing the tumor-associated antigen Lewis Y It represents an attractive target for cancer immunotherapy and can be used in treatment of breast cancer MW 146 38 KD