Learn More
Biotium MB succinimidyl ester
Catalog No. NC1652689
Encompass_Preferred
The amine reactive succinimidyl ester of MB (methylene blue) can be conjugated to peptides, proteins, drugs, polymeric materials and biomolecules with primary amine groups.  The conjugate will have a blue color and be able to complex with nucleic acids.  Methylene blue is a well known redox dye which can be reversibly reduced to its colorless leuco form.  Upon oxidation, the blue color of the dye is recovered.
Catalog No. NC1652689 Supplier Biotium Supplier No. 40075
The amine reactive succinimidyl ester of MB (methylene blue) can be conjugated to peptides, proteins, drugs, polymeric materials and biomolecules with primary amine groups.  The conjugate will have a blue color and be able to complex with nucleic acids.  Methylene blue is a well known redox dye which can be reversibly reduced to its colorless leuco form.  Upon oxidation, the blue color of the dye is recovered.