Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec MB100 Cup Sink Grossing Station Countertop
SDP

Catalog No. 22444194 Shop All Mopec Products
Change view
Click to view available options
Height (Metric):
116 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric)
22-444-194 116 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-444-194 Supplier Mopec Supplier No. MB100

missing translation for 'validMainframeMsg'

Ideal for laboratories with space constraints. Mopec™ MB100 Cup Sink Grossing Station Countertop is a compact workstation that rests on a counter top and saves valuable floor space.

  • Includes hot/cold water fixture, wrist blade handles with swing spout faucet and built-in vacuum breaker
  • Removable polyethylene cutting board
  • Washable ruler with inch and centimeter markings
  • Ruler is located on the inside lip of the workstation
  • Magnetic tool bar holds instruments when not in use
  • One large and one small stainless steel organizer bin
  • Stainless steel c-fold paper towel dispenser
  • Two energy saving, long life LED lights
  • Back draft ventilation designed to safely and efficiently remove formaldehyde fumes and hazardous chemical vapors
  • Equipped with a factory installed exhaust duct which can be connected to any in-house ventilation system
  • All plumbing and electrical work accomplished at factory
  • Weight: 750 lb.; Shipping Weight: 850 lb.

Warranty and Services

1 Year Warranty

Specifications

Description Grossing Station - Countertop, Cup Sink
Electrical Requirements GFCI electrical receptacle, 1 circuit, 15 amps
Frequency 60 Hz
Voltage 115 V
Ventilation Back draft ventilation
Depth (Metric) 122 cm
Depth (Metric) Sink 7.6 cm
Height (Metric) 116 cm
Length (Metric) Sink 25.4 cm
Weight (English) 750 lb.
Width (Metric) 74 cm
Width (Metric) Sink 12.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.