Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec MB200 Large Sink Countertop Grossing Station
SDP

Catalog No. 22444196 Shop All Mopec Products
Change view
Click to view available options
Height (Metric):
115.6cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric)
22-444-196 115.6cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-444-196 Supplier Mopec Supplier No. MB200

missing translation for 'validMainframeMsg'

Designed to maximize floor space, this grossing station is a compact counter top workstation featuring one of the deepest sinks in the industry. Mopec™ MB200 Large Sink Countertop Grossing Station is an integrally constructed sink that offers limitless utility for the any size lab.

  • All stainless steel construction
  • Large deep sink
  • Includes hot/cold water fixture
  • Wrist blade handles with swing spout faucet and built-in vacuum breaker
  • Recessed stainless steel work surface with removable polyethylene cutting board
  • One large and one small stainless steel organizer bin
  • Washable ruler with inch/centimeter markings
  • Ruler is located on the inside lip of the workstation
  • Magnetic tool bar holds instruments when not in use
  • Stainless steel c-fold paper towel dispenser
  • Two energy saving, long life LED lights
  • GFCI Electrical receptacle
  • Back draft ventilation designed to safely and efficiently remove formaldehyde fumes and hazardous chemical vapors
  • Equipped with a factory installed exhaust duct which can be connected to any in-house ventilation system
  • All plumbing and electrical work accomplished at factory
  • Only trade hook-up required

Warranty and Services

1 Year Warranty

Specifications

Product Type Grossing Station
Description Grossing Station
Frequency 60 Hz
Voltage 115 V
Ventilation Back Draft Ventilation
Depth (Metric) 121.9 cm
Depth (Metric) Sink 25 cm
Height (Metric) 115.6cm
Length (Metric) Sink 48cm
Width (Metric) 73.7 cm
Width (Metric) Sink 36 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.