Learn More
Cayman Chemical MMB-4en-PICA butanoic acid metabolite, Purity: ≥98%, 5 mg
SDP
Catalog No. 502991433
Encompass_Preferred
An analytical reference standard that is structurally similar to known synthetic cannabinoids; intended for research and forensic applications
Catalog No. 50-299-1433 Supplier Cayman Chemical Supplier No. 312755 MG
An analytical reference standard that is structurally similar to known synthetic cannabinoids; intended for research and forensic applications